Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nme1, Mouse, Clone: KM1103, Abnova™

Catalog No. 89111423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
35 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-423 35 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-423 Supplier Abnova Corporation Supplier No. MAB3443
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Nme1.

Specifications

Antigen Nme1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone KM1103
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Nme1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Nme1
Gene Symbols Nme1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to rat Nme1.
Quantity 35 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 191575
Test Specificity Reacts with rat NDP kinase beta isoform and human NDP Kinase A (nm23-H1).
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.