Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOP2, Mouse, Clone: 22G1, Abnova™
Mouse monoclonal antibody raised against native NOP2.
Supplier: Abnova Corporation MAB3341
Specifications
NOP2 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:500-1:5000) Immunocytochemistry (1:250-1:2500) Immunofluorescence (1:250-1:2500) The optimal working dilution should be determined by the end user. | |
NOP2 | |
NOP2 | |
Native purified NOP2 from yeast nuclear protein preparations. | |
RUO | |
855664 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
22G1 | |
Mouse monoclonal antibody raised against native NOP2. | |
In PBS (50% glycerol, 0.09% sodium azide) | |
YNA1 | |
Mouse | |
200 μL | |
Primary | |
Yeast | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction