Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-NOS1AP, Clone: 6E1.1,

Catalog No. MABN229MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN229MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN229MI Supplier MilliporeSigma™ Supplier No. MABN229
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects NOS1AP clone: 6E1.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase(nNOS) protein or CAPON (UniProt O75052), also known as Nitric oxide synthase (NOS) 1 adaptor protein, C-terminal PDZ ligand of nNOS, C-terminal PDZ domain ligand of nNOS, Ligand of nNOS with carboxyl-terminal PDZ domain) is a cytosolic protein encoded by the NOS1AP gene (Entrez Gene ID 9722) in human. NOS1AP protein (CAPON) contains a C-terminal PDZ-binding domain and an N-terminal phosphotyrosine binding (PTB) domain that mediates its interactions with nNOS and the small monomeric G protein Dexras1, respectively. Originally it was characterized as a competitor against PSD95 for nNOS interaction, but later evidence indicates that nNOS-CAPON association contributes to the modulation of anxiety-related behavior via regulating Dexras1-ERK signaling. CAPON is also reported to co-localize with the L-type calcium channel and the inwardly rectifying potassium channel Kir3.1 in murine cardiac myocytes. In addition, CAPON is significantly upregulated in mice with dystrophic cardiomyopathy, implicating a role of NOS1AP protein on cardiac arrhythmias, most likely through S-nitrosylation of L-type calcium channel and potassium channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOS1AP
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6E1.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O75052
Gene Symbols NOS1AP; CAPON; KIAA0464
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the C-terminal of human NOS1AP.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_055512
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.