Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nos3, Mouse, Clone: M221, Abnova™

Catalog No. 89110405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-405 Supplier Abnova Corporation Supplier No. MAB1390
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Nos3.

Specifications

Antigen Nos3
Applications ELISA, Western Blot
Classification Monoclonal
Clone M221
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Nos3.
Dilution ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene Nos3
Gene Alias 2310065A03Rik/Nos-3/eNOS/ecNOS
Gene Symbols Nos3
Host Species Mouse
Immunogen Recombinant protein corresponding to C terminal of mouse Nos3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18127
Test Specificity This sequence is conserved in human and rat eNOS, and has low homology to other NOS family members.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.