Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NP, Mouse, Clone: 33s6, Abnova™

Catalog No. 89113650 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-650 Supplier Abnova Corporation Supplier No. MAB8927
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native NP.

Specifications

Antigen NP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 33s6
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native NP.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene NP
Gene Symbols NP
Host Species Mouse
Immunogen Native bacterin consisting of H5N1 and H9N2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3654618
Test Specificity This antibody recognizes Influenza A virus NP.
Target Species Virus
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.