Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPAS2, Mouse, Clone: 6C9, Abnova™

Catalog No. 89105021 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-021 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-021 Supplier Abnova Corporation Supplier No. H00004862M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NPAS2.

The protein encoded by this gene is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. A similar mouse protein may play a regulatory role in the acquisition of specific types of memory. It also may function as a part of a molecular clock operative in the mammalian forebrain. [provided by RefSeq

Sequence: NLTTPASTSQDASQCQPSPDFSHDRQLRLLLSQPIQPMMPGSCDARQPSEVSRTGRQVKYAQSQTVFQNPDAHPANSSSAPMPVLLMGQAVLH

Specifications

Antigen NPAS2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6C9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NPAS2.
Formulation ascites with no preservative
Gene NPAS2
Gene Accession No. NM_002518
Gene Alias FLJ23138/MGC71151/MOP4/PASD4/bHLHe9
Gene Symbols NPAS2
Host Species Mouse
Immunogen NPAS2 (NP_002509,646 a.a. ∽ 738 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4862
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.