Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPPB, Mouse, Clone: 3A6F7C7, Abnova™

Catalog No. 89108698 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-698 Supplier Abnova Corporation Supplier No. MAB10511
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of NPPB.

This gene is a member of the natriuretic peptide family and encodes a secreted protein which functions as a cardiac hormone. The protein undergoes two cleavage events, one within the cell and a second after secretion into the blood. The protein's biological actions include natriuresis, diuresis, vasorelaxation, inhibition of renin and aldosterone secretion, and a key role in cardiovascular homeostasis. A high concentration of this protein in the bloodstream is indicative of heart failure. Mutations in this gene have been associated with postmenopausal osteoporosis. [provided by RefSeq]

Sequence: EPLQESPRPTGVWC

Specifications

Antigen NPPB
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 3A6F7C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of NPPB.
Dilution ELISA (1:10000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene NPPB
Gene Alias BNP
Gene Symbols NPPB
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human NPPB.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4879
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.