Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-NQO2, Clone: 4A4.1,

Catalog No. MABC1038 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1038 Supplier MilliporeSigma™ Supplier No. MABC1038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects NQO2 clone: 4A4.1 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

Ribosyldihydronicotinamide dehydrogenase [quinone] (EC 1.10.5.1; UniProt P16083; also known as NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, QR2) is encoded by the NQO2 (also known as NMOR2) gene (Gene ID 4835) in human. NQO1 (EC 1.6.5.2) and NQO2 (EC 1.10.5.1; formerly EC 1.10.99.2) constitute the quinone oxidoreductase family in human. NQO1 is involved in vitamin K metabolism and in reducing the cellular quinone concentration to prevent the build-up of cellular reactive oxygen species (ROS). Both enzymes contain a tightly bound FAD as the oxidant cofactor, but differ in their reductant cofactors utilization. While NQO1 can efficiently utilize NADH or NADPH, NQO2 exhibits only low activity in the presence of NADH or NADPH. Instead, non-physiological compounds, such as dihydronicotinamide riboside (NRH), N-methyldihydronicotinamide, and dihydrobenzylnicotamide, are reported to act as good reductant cofactors in NQD2-catalyzed reactions in vitro. NQO1 & NQO2 catalyzed the reduction of a wide variety of substrates, including quinones, nitro-compounds, and iron (III) ions. NQO1 is inhibited by the anticoagulant dicoumarol, while NQO2 is only weakly inhibited by dicoumarol and more strongly inhibited by resveratrol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NQO2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4A4.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P16083
Gene Symbols NQO2; NMOR2
Host Species Mouse
Immunogen GST-tagged recombinant human NQO2 N-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_000895
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.