Learn More
NR4A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NR4A3.
Supplier: Abnova Corporation H00008013A01
Description
This gene encodes a member of the steroid-thyroid hormone-retinoid receptor superfamily. The encoded protein may act as a transcriptional activator. The protein can efficiently bind the NGFI-B Response Element (NBRE). Three different versions of extraskeletal myxoid chondrosarcomas (EMCs) are the result of reciprocal translocations between this gene and other genes. The translocation breakpoints are associated with Nuclear Receptor Subfamily 4, Group A, Member 3 (on chromosome 9) and either Ewing Sarcome Breakpoint Region 1 (on chromosome 22), RNA Polymerase II, TATA Box-Binding Protein-Associated Factor, 68kDa (on chromosome 17), or Transcription factor 12 (on chromosome 15). Four transcript variants encoding three distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: LDYSRYCPTDQAAAGTDAEHVQQFYNLLTASIDVSRSWAEKIPGFTDLPKEDQTLLIESAFLELFVLRLSIRSNTAEDKFVFCNGLVLHRLQCLRGFGEWLDSIKDFSSpecifications
NR4A3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NR4A3. | |
NR4A3 | |
CHN/CSMF/MINOR/NOR1/TEC | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006981 | |
NR4A3 | |
NR4A3 (NP_008912, 414 a.a. ∽ 521 a.a) partial recombinant protein with GST tag. | |
RUO | |
8013 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only