Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRCAM, Mouse, Clone: RNL-1, Abnova™

Catalog No. 89111470 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-470 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-470 Supplier Abnova Corporation Supplier No. MAB3561
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native NRCAM.

Cell adhesion molecules (CAMs) are members of the immunoglobulin superfamily. This gene encodes a neuronal cell adhesion molecule with multiple immunoglobulin-like C2-type domains and fibronectin type-III domains. This ankyrin-binding protein is involved in neuron-neuron adhesion and promotes directional signaling during axonal cone growth. This gene is also expressed in non-neural tissues and may play a general role in cell-cell communication via signaling from its intracellular domain to the actin cytoskeleton during directional cell migration. Allelic variants of this gene have been associated with autism and addiction vulnerability. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen NRCAM
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RNL-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native NRCAM.
Dilution Immunohistochemistry (1:10-1:25) The optimal working dilution should be determined by the end user.
Formulation In cell culture supernatant (0.09% sodium azide)
Gene NRCAM
Gene Alias KIAA0343/MGC138845/MGC138846
Gene Symbols NRCAM
Host Species Mouse
Immunogen Native purified NRCAM from NCI-H82 cell.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4897
Test Specificity This antibody reacts with extracellular domain of the NRCAM.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.