Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPB1, Mouse, Clone: 7D5, Abnova™

Catalog No. 89110693 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-693 Supplier Abnova Corporation Supplier No. MAB1890

Mouse monoclonal antibody raised against partial recombinant Nrg1.

Specifications

Antigen HSPB1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7D5
Conjugate Unconjugated
Description neuregulin 1
Formulation In PBS (0.08% sodium azide)
Gene Nrg1
Gene Symbols Nrg1
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular region of rat Nrg1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112400
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.