Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF, Mouse, Clone: NSF-1, Abnova™

Catalog No. 89110735 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-735 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-735 Supplier Abnova Corporation Supplier No. MAB1966

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against recombinant NSF.

Specifications

Antigen NSF
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone NSF-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant NSF.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 150mM NaCl, pH 7.5 (50% glycerol, 3mM sodium azide)
Gene NSF
Gene Alias SKD2
Gene Symbols NSF
Host Species Mouse
Immunogen Recombinant protein of human NSF.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4905
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.