Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSMAF, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105854 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-854 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-854 Supplier Abnova Corporation Supplier No. H00008439A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NSMAF.

This gene encodes a WD-repeat protein that binds the cytoplasmic sphingomyelinase activation domain of the 55kD tumor necrosis factor receptor. This protein is required for TNF-mediated activation of neutral sphingomyelinase and may play a role in regulating TNF-induced cellular responses such as inflammation. Alternative splicing results in multiple transcript variants

Sequence: RHVLSTGTDGCLNVIDVQTGMLISSMTSDEPQRCFVWDGNSVLSGSQSGELLVWDLLGAKISERIQGHTGAVTCIWMNEQCSSIITGGEDRQIIFWKLQY

Specifications

Antigen NSMAF
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NSMAF.
Formulation 50% glycerol
Gene NSMAF
Gene Accession No. NM_003580
Gene Alias FAN
Gene Symbols NSMAF
Host Species Mouse
Immunogen NSMAF (NP_003571, 818 a.a. ∽ 917 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8439
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.