Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-NTR1, Clone: 8F11.1,

Catalog No. MABN350MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN350MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN350MI Supplier MilliporeSigma™ Supplier No. MABN350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects NTR1 clone: 8F11.1 in Human, Mouse samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Neurotensin receptor type 1 (UniProt P30989; also known as High-affinity levocabastine-insensitive neurotensin receptor, NT-R-1, NTR1, NTRH) is encoded by the NTSR1 (also known as NTRR) gene (Gene ID 4923) in human. Neurotensin (NT) is a tridecapeptide that acts as a neuromodulator in the CNS. It also behaves as a hormone in the gastrointestinal tract. Four different NTSRs have been reported. NTSR1 and NTSR2 are seven-transmembrane G-protein-coupled receptors (GPCRs), while NTSR3 and NTSR4 are members of the sortilin protein family with a single-transmembrane spanning domain. NTSR1 is a high affinity NT receptor and plays an important role in cancer cell proliferation, survival, mobility, and invasion. NT has also been implicated in the pathophysiology of several CNS disorders, such as schizophrenia, drug abuse, eating disorders, and Parkinson’s disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8F11.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P30989
Gene Symbols NTSR1; NTRR
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human NTR1 near the C-terminus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_002522
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.