Learn More
NTRK3, Mouse, Clone: 4G5, Abnova™
Mouse monoclonal antibody raised against partial recombinant NTRK3.
Supplier: Abnova Corporation MAB10587
Description
This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position. Mutations in this gene have been associated with medulloblastomas, secretory breast carcinomas and other cancers. [provided by RefSeq
Specifications
NTRK3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
NTRK3 | |
NTRK3 | |
Recombinant hIgGFc tag fusion protein corresponding to extracellular fragment at amino acids 32-429 of human NTRK3. | |
RUO | |
4916 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
4G5 | |
Mouse monoclonal antibody raised against partial recombinant NTRK3. | |
In ascites (0.03% sodium azide) | |
TRKC/gp145(trkC) | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.