Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTRK3, Mouse, Clone: 7H6, Abnova™

Catalog No. 89108778 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-778 Supplier Abnova Corporation Supplier No. MAB10586
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NTRK3.

This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position. Mutations in this gene have been associated with medulloblastomas, secretory breast carcinomas and other cancers. [provided by RefSeq

Specifications

Antigen NTRK3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7H6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NTRK3.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene NTRK3
Gene Alias TRKC/gp145(trkC)
Gene Symbols NTRK3
Host Species Mouse
Immunogen Recombinant hIgGFc tag fusion protein corresponding to extracellular fragment at amino acids 32-429 of human NTRK3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4916
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.