Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclei, Mouse, Clone: 235-1, Abnova™

Catalog No. 89136428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-428 Supplier Abnova Corporation Supplier No. MAB8178
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Nuclei.

Specifications

Antigen Nuclei
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 235-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Nuclei.
Dilution Immunofluorescence (1:100) Immunohistochemistry (1: 100) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Native puridfied from human cell homogenate.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody has been directly tested for reactivity in human. This antibody does not react with mouse or rat nuclei. specific to human nuclei. Stains nuclei of all human cell types giving a diffuse nuclear staining pattern. Chromosomes are negatively stained in metaphase cells. By immunoprecipitation This antibody reacts with an 80 KDa and 70 KDa band. This antibody works on acetone fixed and paraformaldehyde fixed cells. It is suggested that you permeabilize the cells with 0.1-0.4% Triton X100. This antibody also works for immunohistochemistry on paraformaldehyde fixed cryostat sections.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.