Learn More
NUP133, Mouse, Clone: 3E8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NUP133.
Supplier: Abnova Corporation H00055746M01A
Description
The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. The nucleoporin protein encoded by this gene displays evolutionarily conserved interactions with other nucleoporins. This protein, which localizes to both sides of the nuclear pore complex at interphase, remains associated with the complex during mitosis and is targeted at early stages to the reforming nuclear envelope. This protein also localizes to kinetochores of mitotic cells. [provided by RefSeq
Sequence: LKLEILCKALQRDNWSSSDGKDDPIEVSKDSIFVKILQKLLKDGIQLSEYLPEVKDLLQADQLGSLKSNPYFEFVLKANYEYYVQGQSpecifications
NUP133 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_018230 | |
NUP133 | |
NUP133 (NP_060700,1069 a.a. ∽ 1155 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
55746 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3E8 | |
Mouse monoclonal antibody raised against a partial recombinant NUP133. | |
NUP133 | |
FLJ10814/MGC21133/hNUP133 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.