Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP133, Mouse, Clone: 4F18, Abnova™

Catalog No. 89107065 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-065 Supplier Abnova Corporation Supplier No. H00055746M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NUP133.

The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. The nucleoporin protein encoded by this gene displays evolutionarily conserved interactions with other nucleoporins. This protein, which localizes to both sides of the nuclear pore complex at interphase, remains associated with the complex during mitosis and is targeted at early stages to the reforming nuclear envelope. This protein also localizes to kinetochores of mitotic cells. [provided by RefSeq

Sequence: LKLEILCKALQRDNWSSSDGKDDPIEVSKDSIFVKILQKLLKDGIQLSEYLPEVKDLLQADQLGSLKSNPYFEFVLKANYEYYVQGQ

Specifications

Antigen NUP133
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F18
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NUP133.
Formulation PBS with no preservative; pH 7.4
Gene NUP133
Gene Accession No. NM_018230
Gene Alias FLJ10814/MGC21133/hNUP133
Gene Symbols NUP133
Host Species Mouse
Immunogen NUP133 (NP_060700,1069 a.a. ∽ 1155 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55746
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.