Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXF2, Mouse, Clone: 4F5-3A2, Abnova™

Catalog No. 89107098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-098 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-098 Supplier Abnova Corporation Supplier No. H00056001M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NXF2.

This gene is one of a family of nuclear RNA export factor genes. It encodes a protein that is involved in mRNA export, is located in the nucleoplasm, and is associated with the nuclear envelope. Alternative splicing seems to be a common mechanism in this gene family. Two variants have been found for this gene. [provided by RefSeq

Sequence: MCSTLKKCGTYRTEVAECHDHGSTFQGRKKGGSSFRDNFDKRSCHYEHGGYERPPSHCQENDGSVEVRDVHKDQQLRHTPYSIRCERRMKWHSEDEIRITTWRNRKPPERKMGQNTQDGYTRNWFKVTIPYGIKYDKAWLMNSIQSHCSDRFTPVDFHYVRNRACFFVQDASAASALKDVSYKIYDDENQKICIFVNHSTAPYSVKNKLRPGQMEMLKLTMNKRYNVSQQALDLQNLRFDPDLMGRDIDIILNRRNCMAATLKIIERNFPELLSLNLCNNKLYQLDGLSDITEKAPKVKTLNLSKNKLESAWELGKVKGLKLEELWLEGNPLCSTFSDQSAYVSAIRDCFPKLLRLDGRELSAPVIVDIDSSETMKPCKENFTGSETLKHLVLQFLQQYYSIYDSGDRQGLLGAYHDEACFSLAIPFDPKDSAPSSLCKYFEDSRNMKTLKDPYLKGELLRRTKRDIVDSLSALPKTQHDLSSILVDVWCQTERMLCFSVNGVFKEVEGQSQGSVLAFTRTFIATPGSSSSLCIVNDELFVRDASPQETQSAFSIPVSTLSSSSEPSLSQEQQEMVQAFSAQSGMKLEWSQKCLQDNEWNYTRAGQAFTMLQTEGKIPAEAFKQIS

Specifications

Antigen NXF2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F5-3A2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant NXF2.
Formulation ascites with no preservative
Gene NXF2
Gene Accession No. BC015020
Gene Alias FLJ20416/TAPL-2
Gene Symbols NXF2
Host Species Mouse
Immunogen NXF2 (AAH15020,1 a.a. ∽ 626 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56001
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.