Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXF5, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107096 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-096 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-096 Supplier Abnova Corporation Supplier No. H00055998A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NXF5.

This gene is one member of a family of nuclear RNA export factor genes. The encoded protein can bind RNA, and is implicated in mRNA nuclear export. However, this protein has lost several C-terminal protein domains found in other family members that are required for export activity, and may be an evolving pseudogene. Alternatively spliced transcript variants have been described, but most are candidates for nonsense-mediated decay (NMD) and may not express proteins in vivo. [provided by RefSeq

Sequence: FTGSETLKHLVLQFLQQSNLCKYFKDSRNIKILKDPYLQRKLLKHTKCPRNVDSLSALPETQHDFTSILVDMWYQTVNTCFLPRAGPESQRWWCLLSLKW

Specifications

Antigen NXF5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NXF5.
Formulation 50% glycerol
Gene NXF5
Gene Accession No. NM_032946
Gene Symbols NXF5
Host Species Mouse
Immunogen NXF5 (NP_116564, 271 a.a. ∽ 370 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55998
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.