Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O3FAR1, Mouse, Clone: 2B6, Abnova™

Catalog No. 89110596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-596 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-596 Supplier Abnova Corporation Supplier No. MAB1725
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of O3FAR1.

GPR120 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) (Fredriksson et al., 2003 [PubMed 14623098]).[supplied by OMIM

Specifications

Antigen O3FAR1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 2B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of O3FAR1.
Dilution Immunoprecipitation (5 ug/mL) Flow Cyt (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2.0% Block Ace)
Gene O3FAR1
Gene Alias BMIQ10/GPR120/GPR129/GT01/PGR4
Gene Symbols O3FAR1
Host Species Mouse
Immunogen A synthetic peptide corresponding to human O3FAR1.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338557
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.