Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OAT, Mouse, Clone: AT23A2, Abnova™

Catalog No. 89113467 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-467 Supplier Abnova Corporation Supplier No. MAB8480
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant OAT.

This gene encodes the mitochondrial enzyme ornithine aminotransferase, which is a key enzyme is the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA. Mutations that result in a deficiency of this enzyme cause the autosomal recessive eye disease Gyrate Atrophy. [provided by RefSeq

Specifications

Antigen OAT
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AT23A2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant OAT.
Dilution Western Blot (1:250-1:500) Immunofluorescence (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene OAT
Gene Alias DKFZp781A11155/HOGA
Gene Symbols OAT
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 33-439 of human OAT.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4942
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.