Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OBSCN, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107426 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-426 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-426 Supplier Abnova Corporation Supplier No. H00084033A02
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant OBSCN.

The obscurin gene spans more than 150kb, contains over 80 exons and encodes a protein of approximately 720kDa. The encoded protein contains 68 Ig domains, 2 fibronectin domains, 1 calcium/calmodulin-binding domain, 1 RhoGEF domain with an associated PH domain, and 2 serine-threonine kinase domains. This protein belongs to the family of giant sacromeric signaling proteins that includes titin and nebulin, and may have a role in the organization of myofibrils during assembly and may mediate interactions between the sarcoplasmic reticulum and myofibrils. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: KAGMGPYSSPSEQVLLGGPSHLASEEESQGRSAQPLPSTKTFAFQTQIQRGRFSVVRQCWEKASGRALAAKIIPYHPKDKTAVLREYEALKGLRHPHLA

Specifications

Antigen OBSCN
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant OBSCN.
Formulation 50% glycerol
Gene OBSCN
Gene Accession No. XM_290923
Gene Alias DKFZp666E245/FLJ14124/KIAA1556/KIAA1639/MGC120409/MGC120410/MGC120411/MGC120412/MGC138590/UNC89
Gene Symbols OBSCN
Host Species Mouse
Immunogen OBSCN (XP_290923, 1551 a.a. ∽ 1649 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84033
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.