Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Oct-1 (Ascites Free), Clone: YL15,

Catalog No. MAB5434C Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5434C 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5434C Supplier MilliporeSigma™ Supplier No. MAB5434C
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Oct-1 (Ascites Free) clone: YL15 in Human, Mouse samples, and it is validated for ChIP, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting

POU domain, class 2, transcription factor 1 (UniProt P14859; also known as NF-A1, Oct-1, Octamer-binding protein 1, Octamer-binding transcription factor 1, OTF-1) is encoded by the POU2F1 (also known as OCT1, OTF1) gene (Gene ID 5451) in human. Oct-1 belongs to the family of octamer transcription factors characterized by their binding to the ATTTGCAT octamer sequence via their POU domain (a.a. 280-354 of human Oct-1). Oct1 is widely expressed in adult tissues, however, Oct-1 knockdown has little impact on cell growth and viability in culture. Oct-1 controls multiple stem cell phenotypes in normal and tumor cells. Oct-1 promotes resistance to genotoxic and oxidative stresses in stem cells. Loss of Oct-1 antagonizes oncogenic transformation in vitro and tumorigenicity in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-1 (Ascites Free)
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone YL15
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P14859
Gene Symbols POU2F1; OCT1; OTF1
Host Species Mouse
Immunogen Recombinant full-length human Oct-1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001185712
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.