Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODZ1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106235 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-235 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-235 Supplier Abnova Corporation Supplier No. H00010178A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ODZ1.

The protein encoded by this gene belongs to the tenascin family and teneurin subfamily. It is expressed in the neurons and may function as a cellular signal transducer. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: TRRFADIQLQHGALCFNIRYGTTVEEEKNHVLEIARQRAVAQAWTKEQRRLQEGEEGIRAWTEGEKQQLLSTGRVQGYDGYFVLSVEQYLELSDSANNIHFMRQSEIGRR

Specifications

Antigen ODZ1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ODZ1.
Formulation 50% glycerol
Gene ODZ1
Gene Accession No. NM_014253
Gene Alias ODZ3/TEN-M1/TNM/TNM1
Gene Symbols ODZ1
Host Species Mouse
Immunogen ODZ1 (NP_055068, 2616 a.a. ∽ 2725 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10178
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.