Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLR1, Mouse, Clone: LOX20-7, Abnova™

Catalog No. 89111223 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-223 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-223 Supplier Abnova Corporation Supplier No. MAB3039
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant OLR1.

This gene encodes a receptor protein which belongs to the C-type lectin superfamily. This gene is regulated through the cyclic AMP signaling pathway. The encoded protein binds, internalizes and degrades oxidized low-density lipoprotein. This protein may be involved in the regulation of Fas-induced apoptosis. This protein may play a role as a scavenger receptor. Mutations of this gene have been associated with atherosclerosis, risk of myocardial infarction, and may modify the risk of Alzheimer's disease. Alternative transcript variants have been described but their full-length sequence has not been determined. [provided by RefSeq]

Specifications

Antigen OLR1
Applications ELISA, Western Blot
Classification Monoclonal
Clone LOX20-7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant OLR1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene OLR1
Gene Alias CLEC8A/LOX1/SCARE1
Gene Symbols OLR1
Host Species Mouse
Immunogen Recombinant protein corresponding to human OLR1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4973
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.