Learn More
OPRL1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant OPRL1.
Supplier: Abnova Corporation H00004987A01
Description
The protein encoded by this gene is a G protein-coupled receptor whose expression can be induced by phytohemagglutinin. The encoded integral membrane protein is a receptor for the 17 aa neuropeptide nociceptin/orphanin FQ. This gene may be involved in the regulation of numerous brain activities, particularly instinctive and emotional behaviors. A promoter for this gene also functions as a promoter for another gene, regulator of G-protein signalling 19 (RGS19), located on the opposite strand. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: DILLGFWPFGNALCKTVIAIDYYNMFTSTFTLTAMSVDRYVAICHPIRALDVRTSSKAQAVNVAIWALASVVGVPVAIMGSAQVEDEEIECLVEIPTPQDYWGSpecifications
OPRL1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant OPRL1. | |
OPRL1 | |
KOR-3/MGC34578/NOCIR/OOR/ORL1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC038433 | |
OPRL1 | |
OPRL1 (AAH38433, 110 a.a. ∽ 212 a.a) partial recombinant protein with GST tag. | |
RUO | |
4987 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only