Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORAI1, Mouse, Clone: 6D11A11, Abnova™

Catalog No. 89108319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-319 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-319 Supplier Abnova Corporation Supplier No. MAB10004
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of ORAI1.

CRACM1 is a plasma membrane protein essential for store-operated calcium entry (Vig et al., 2006 [PubMed 16645049]).[supplied by OMIM

Specifications

Antigen ORAI1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6D11A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of ORAI1.
Dilution Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ORAI1
Gene Alias CRACM1/FLJ14466/ORAT1/TMEM142A
Gene Symbols ORAI1
Host Species Mouse
Immunogen A synthetic peptide corresponding to 16 amino acids at C-terminus of human ORAI1.
Purification Method IgG purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84876
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.