Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORAI3, Mouse, Clone: 2H2G9, Abnova™

Catalog No. 89113340 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-340 Supplier Abnova Corporation Supplier No. MAB8084
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against synthetic peptide of ORAI3.

Specifications

Antigen ORAI3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2H2G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of ORAI3.
Dilution Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ORAI3
Gene Alias MGC13024/TMEM142C
Gene Symbols ORAI3
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus 19 amino acids of human ORAI3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93129
Target Species Human, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.