Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSR1, Mouse, Clone: 4B12, Abnova™

Catalog No. 89107605 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-605 Supplier Abnova Corporation Supplier No. H00130497M08J
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant OSR1.

Sequence: MGSKTLPAPVPIHPSLQLTNYSFLQAVNGLPTVPSDHLPNLYGFSALHAVHLHQWTLGYPAMHLPRSSFSKVPGTVSSLVDARFQLPAFPWFPHVIQPKP

Specifications

Antigen OSR1
Applications ELISA
Classification Monoclonal
Clone 4B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant OSR1.
Formulation PBS with no preservative; pH 7.4
Gene OSR1
Gene Accession No. NM_145260
Gene Alias ODD
Gene Symbols OSR1
Host Species Mouse
Immunogen OSR1 (NP_660303,1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130497
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.