Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-oxDJ-1 (Cys106), Clone: M149,

Catalog No. MABN1773MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1773MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1773MI Supplier MilliporeSigma™ Supplier No. MABN1773
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects oxDJ-1 (Cys106) clone: M149 in Human, Mouse samples, and it is validated for Immunohistochemistry, Western Blotting

Protein DJ-1 (UniProt Q99497; also known as Parkinson disease (autosomal recessive, early onset) protein 7, Epididymis secretory sperm binding protein Li 67p, Oncogene DJ1) is encoded by the PARK7 (also known as DJ1, HEL-S-67p) gene (Gene ID 11315) in human. DJ-1 belongs to the ThiJ/PfpI family of proteins characterized by a ThiJ domain. DJ-1 is reported to be involved in multiple physiological processes, including cellular oncogenic transformation, RNA-protein interaction regulation, sperm fertilization, antioxidative defense, and transcriptional regulation. Mutations in the DJ-1 gene, PARK7, are known causes of autosomal recessive parkinsonism with a clinical presentation similar to other forms of recessive PD syndromes. DJ-1 cysteine residue 106 (Cys106) is preferentially oxidized through direct oxygen addition in response to cellular oxidative stress. Cysteine forms 3 different oxidized species, cysteine–sulfenic acid (Cys-SOH), cysteine–sulfinic acid (Cys-SO2H), and cysteine–sulfonic acid (Cys-SO3H). The Cys-SO2H form of oxDJ-1 is likely the active form, and further oxidation to Cys-SO3H leads to loss of biologic function. The levels of oxDJ-1 in erythrocytes from unmedicated PD patients are reported to be markedly higher than those from medicated PD patients and healthy subjects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen oxDJ-1 (Cys106)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone M149
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q99497
Gene Symbols PARK7
Host Species Mouse
Immunogen His-tagged oxidized recombinant DJ-1 purified from H2O2-treated E. coli host corresponding to the Cys106-SO2H/-SO3H of human oxDJ-1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001116849
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.