Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-P-Selectin/CD62P, Clone: AK-4, PE, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Specifications

Antigen P-Selectin/CD62P
Clone AK-4
Concentration 0.2 mg/mL
Dilution Flow Cytometry 1:10-1:1000
Applications Flow Cytometry
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP143850
SDP
View Documents
Novus Biologicals
NBP143850100TESTS
100 Tests N/A N/A N/A
Description

Description

P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

P-Selectin/CD62P
0.2 mg/mL
Flow Cytometry
PE
RUO
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
6403
The immunogen for this antibody was CD62P.
Primary
Store at 4C. Do not freeze.
AK-4
Flow Cytometry 1:10-1:1000
Monoclonal
Mouse
Human
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
SELP
IgG1 κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.