Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106494 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-494 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-494 Supplier Abnova Corporation Supplier No. H00011240A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PADI2.

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type II enzyme is the most widely expressed family member. Known substrates for this enzyme include myelin basic protein in the central nervous system and vimentin in skeletal muscle and macrophages. This enzyme is thought to play a role in the onset and progression of neurodegenerative human disorders, including Alzheimer disease and multiple sclerosis, and it has also been implicated in glaucoma pathogenesis. This gene exists in a cluster with four other paralogous genes. [provided by RefSeq]

Sequence: MLRERTVRLQYGSRVEAVYVLGTYLWTDVYSAAPAGAQTFSLKHSEHVWVEVVRDGEAEEVATNGKQRWLLSPSTTLRVTMSQASTEASSDKVTVNYYDEEGSIPIDQ

Specifications

Antigen PADI2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PADI2.
Formulation 50% glycerol
Gene PADI2
Gene Accession No. NM_007365
Gene Alias KIAA0994/PAD-H19/PAD2/PDI2
Gene Symbols PADI2
Host Species Mouse
Immunogen PADI2 (NP_003008, 1 a.a. ∽ 108 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11240
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.