Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PAFAH 1B3, Clone: 3E4.1,

Catalog No. MABS1188 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1188 Supplier MilliporeSigma™ Supplier No. MABS1188
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PAFAH 1B3 clone: 3E4.1 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Platelet-activating factor acetylhydrolase IB subunit gamma (EC 3.1.1.47; UniProt Q15102; also known as PAF acetylhydrolase 29kDa subunit, PAF-AH 29kDa subunit, PAF-AH1b subunit alpha 1, PAF-AH subunit gamma, PAFAH subunit gamma) is encoded by the PAFAH1B3 (also known as PAFAHG) gene (Gene ID 5050) in human. Platelet-activating factor (PAF) acetylhydrolase (PAF-AH) negatively regulates PAF potency by catalyzing the removal of the acetyl group at the sn-2 position of the lipid messenger. There are two types of intracellular PAF-AH and a plasma type PAF-AH. Type I intracellular PAF-AH is a heterotrimeric G-protein-like complex composed of alpha1 (originally called gamma), alph2 (originally called alpha), and beta subunits. Both alpha1 and alpha2 contains a catalytic site. The alpha2/2 homodimer hydrolyzes PAF (1-O-alkyl-2-acetyl-sn-glycero-3-PC) and 1-O-alkyl-2-acetyl-sn-glycero-3-PE more efficiently, while the alpha1/1 homodimer and alpha1/2 heterodimer are more efficient toward 1-O-alkyl-2-acetyl-sn-glycero-3-PA than PAF. The alpha 1 subunit is reported to be up-regulated in primary human breast tumors and correlated with poor prognosis. Inactivation of alpha 1 subunit enhances cellular level of tumor-suppressing signaling lipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAFAH 1B3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3E4.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q15102
Gene Symbols PAFAH1B3; PAFAHG
Host Species Mouse
Immunogen GST-tagged recombinant internal fragment from the C-terminal half of human PAFAH1B3.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001139411
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.