Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAFAH1B2, Mouse, Clone: 2F4-1C10, Abnova™

Catalog No. 89105043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-043 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-043 Supplier Abnova Corporation Supplier No. H00005049M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PAFAH1B2.

Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha (MIM 601545), beta, and gamma (MIM 603074) PAFAH subunits.[supplied by OMIM

Sequence: MSQGDSNPAAIPHAAEDIQGDDRWMSQHNRFVLDCKDKEPDVLFVGDSMVQLMQQYEIWRELFSPLHALNFGIGGDTTRHVLWRLKNGELENIKPKVIVVWVGTNNHENTAEEVAGGIEAIVQLINTRQPQAKIIVLGLLPRGEKPNPLRQKNAKVNQLLKVSLPKLANVQLLDTDGGFVHSDGAISCHDMFDFLHLTGGGYAKICKPLHELIMQLLEETPEEKQTTIA

Specifications

Antigen PAFAH1B2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F4-1C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant PAFAH1B2.
Formulation ascites with no preservative
Gene PAFAH1B2
Gene Accession No. BC000398
Gene Symbols PAFAH1B2
Host Species Mouse
Immunogen PAFAH1B2 (AAH00398,1 a.a. ∽ 229 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5049
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.