Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAG1, Mouse, Clone: MEM-255, Abnova™

Catalog No. 89108263 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-263 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-263 Supplier Abnova Corporation Supplier No. MAB0935
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PAG1.

The protein encoded by this gene is a type III transmembrane adaptor protein that binds to the tyrosine kinase csk protein. It is thought to be involved in the regulation of T cell activation. [provided by RefSeq

Specifications

Antigen PAG1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MEM-255
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PAG1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PAG1
Gene Alias CBP/FLJ37858/MGC138364/PAG
Gene Symbols PAG1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 97-432 of human PAG1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55824
Test Specificity This antibody recognizes an epitope (aa 235-280) of Csk-binding protein (Cbp) located in the cytoplasmic domain.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.