Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAIP1, Mouse, Clone: 7E7, Abnova™

Catalog No. 89106367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-367 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-367 Supplier Abnova Corporation Supplier No. H00010605M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PAIP1.

The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified. [provided by RefSeq

Sequence: YPTLSEYVQDFLNHLTEQPGSFETEIEQFAETLNGCVTTDDALQELVELIYQQATSIPNFSYMGARLCNYLSHHLTISPQSGNFRQLLLQRCRTEYEVKDQAAKGDEVTR

Specifications

Antigen PAIP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PAIP1.
Formulation ascites with no preservative
Gene PAIP1
Gene Accession No. NM_182789
Gene Alias MGC12360
Gene Symbols PAIP1
Host Species Mouse
Immunogen PAIP1 (NP_001002,76 a.a. ∽ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10605
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.