Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAIP2, Mouse, Clone: AT1C12, Abnova™

Catalog No. 89109369 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-369 Supplier Abnova Corporation Supplier No. MAB11200
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PAIP2.

Specifications

Antigen PAIP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1C12
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PAIP2.
Dilution Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) with 0.09% sodium azide.
Gene PAIP2
Gene Alias MGC72018/PAIP2A
Gene Symbols PAIP2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-127 of human PAIP2.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51247
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.