Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-pan-AMPA receptor (GluR1-4), Clone: 2D8,

Catalog No. MABN832MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN832MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN832MI Supplier MilliporeSigma™ Supplier No. MABN832
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects pan-AMPA receptor (GluR1-4) clone: 2D8 in Human, Rat samples, and it is validated for Electron Microscopy, ELISA, Enzyme Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting

Glutamate receptors are synaptic receptors located primarily on the membranes of neuronal cells. Upon ligand stimulation, ionotropic glutamate receptors (iGluRs) form non-selective cation channels, while metabotropic glutamate receptors (mGluRs) indirectly activate ion channels via G protein-mediated signaling. AMPA receptors are iGluRs and are also known as alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptors, AMPARs, or quisqualate receptors. AMPARs are composed of four types of subunits, GluR1/GluA1 (UniProt P42261) is encoded by GRIA1, GluR2/GluA2 (UniProt P42262) by GRIA2, GluR3/GluA3 (UniProt P42263) by GRIA3, and GluR4/GluA4 (UniProt P48058) by GRIA4 gene in human. AMPARs are tetrameric in nature, consisting of two sets of homodimers, most often a set of GluR2 dimer is coupled with a set of GluR1, GluR3 or GluR4 homodimer. Anti-pan-AMPA receptor (GluR1-4), clone 2D8, reacts with all four AMPA receptor subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan-AMPA receptor (GluR1-4)
Applications Immunoassay, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2D8
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P42261; P42262; P42263; P48058
Gene Symbols GRIA1; GLUH1; GLUR1
Host Species Mouse
Immunogen Fusion protein coding for the extracellular domain of AMPA Glutamate receptor 1-4 corresponding to human pan-AMPA receptor (GluR1-4).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000818; NP_000817; NP_000819; NP_000820
Target Species Human, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.