Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Pancreatic alpha cell marker, Clone: DHIC2-2C12, Abnova™

Catalog No. 89136323 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-323 Supplier Abnova Corporation Supplier No. MAB7226
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone DHIC2-2C12
Description Mouse monoclonal antibody raised against pancreatic alpha cell marker.
Dilution Flow Cytometry (1:50-1:100) Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant
Immunogen Native purified human pancreatic enriched alpha cells.
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Specific for alpha endocrine cell types.
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.