Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAPPA, Mouse, Clone: 11E4, Abnova™

Catalog No. 89107994 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-994 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-994 Supplier Abnova Corporation Supplier No. MAB0587
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native PAPPA.

This gene encodes a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregnancies with aneuploid fetuses. [provided by RefSeq

Specifications

Antigen PAPPA
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 11E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native PAPPA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene PAPPA
Gene Alias ASBABP2/DIPLA1/IGFBP-4ase/PAPA/PAPP-A/PAPPA1
Gene Symbols PAPPA
Host Species Mouse
Immunogen Native purified human PAPPA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5069
Test Specificity Specific to proMBP subunit.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.