Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP1, Mouse, Clone: 194C1439, Abnova™

Catalog No. 89107867 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-867 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-867 Supplier Abnova Corporation Supplier No. MAB0097
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of PARP1.

This gene encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes. [provided by RefSeq

Specifications

Antigen PARP1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 194C1439
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of PARP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene PARP1
Gene Alias ADPRT/ADPRT1/PARP/PARP-1/PPOL/pADPRT-1
Gene Symbols PARP1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids near 214/215-cleavage site of human PARP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.