Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP3, Mouse, Clone: LA6B10, Abnova™

Catalog No. 89111421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-421 Supplier Abnova Corporation Supplier No. MAB3439

Mouse monoclonal antibody raised against synthetic peptide of PARP3.

The protein encoded by this gene belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This gene encodes the poly(ADP-ribosyl)transferase 3, which is preferentially localized to the daughter centriole throughout the cell cycle. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen PARP3
Applications Immunocytochemistry
Classification Monoclonal
Clone LA6B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of PARP3.
Dilution Immunocytochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In culture supernatant (0.09% sodium azide)
Gene PARP3
Gene Alias ADPRT3/ADPRTL2/ADPRTL3/IRT1/hPARP-3/pADPRT-3
Gene Symbols PARP3
Host Species Mouse
Immunogen A synthetic peptide (conjugated with OVA) corresponding to N-terminus amino acids 8-22 (M8APKPKPWVQTEGPE22) of human PARP3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10039
Test Specificity Recognizes human and mouse PARP-3.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.