Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP4, Mouse, Clone: P193-4, Abnova™

Catalog No. 89135911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-911 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-911 Supplier Abnova Corporation Supplier No. MAB5189
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant PARP4.

Specifications

Antigen PARP4
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone P193-4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant PARP4.
Dilution Immunohistochemistry (Frozen sections) (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 1% BSA, 0.09% sodium azide
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 408-611 of PARP4.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity p193-4 reacts with an internal epitope (amino acids 491-494, HPGE) of the minor vault protein (p193 or VPARP), which is overexpressed in various human non-P-glycoprotein MDR tumor cell lines, accordingly to an increase in the number of vault particles.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.