Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Pax-2, Clone: 4C2.2,

Catalog No. MABE902MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE902MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE902MI Supplier MilliporeSigma™ Supplier No. MABE902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Pax-2 clone: 4C2.2 in Human samples, and it is validated for Western Blotting

Paired box protein Pax-2 (UniProt Q02962; also known as Paired box homeotic gene 2) is encoded by the PAX2 (also known as FSGS7, PAPRS) gene (Gene ID 5076) in human. Pax-2 belongs to the family of transcription factors with conserved DNA-binding paired box domain. The family of paired box (PAX) genes is divided into four subgroups (I–IV). Genes in subgroups II (PAX2, PAX5, and PAX8) and III (PAX3 and PAX7) have been implicated in tumor development. PAX2 is a critical regulator of embryogenesis and it is normally expressed in the pancreas and in subpopulations of nodal lymphocytes in adult tissues. Aberrant PAX2 expression is frequently observed in several cancer types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax-2
Applications Western Blot
Classification Monoclonal
Clone 4C2.2
Formulation Mouse monoclonal IgG2bκ antibody in buffer containing supernatant without preservatives.
Gene Accession No. Q02962
Gene Symbols PAX2
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Pax-2.
Purification Method Unpurified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000269
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Supernatant
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.