Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAXIP1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106543 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-543 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-543 Supplier Abnova Corporation Supplier No. H00022976A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PAXIP1.

This gene is a member of the paired box (PAX) gene family and encodes a nuclear protein with six BRCT (breast cancer carboxy-terminal) domains. This protein plays a critical role in maintaining genome stability, condensation of chromatin and progression through mitosis. [provided by RefSeq

Sequence: HLIASKVTRTVKFLTAISVVKHIVTPEWLEECFRCQKFIDEQNYILRDAEAEVLFSFSLEESLKRAHVSPLFKAKYFYITPGICPSLSTMKAIVECAGGKVLSKQPSF

Specifications

Antigen PAXIP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PAXIP1.
Formulation 50% glycerol
Gene PAXIP1
Gene Accession No. NM_007349
Gene Alias CAGF28/CAGF29/FLJ41049/PACIP1/PAXIP1L/PTIP/TNRC2
Gene Symbols PAXIP1
Host Species Mouse
Immunogen PAXIP1 (NP_620176, 868 a.a. ∽ 975 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22976
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.