Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PC-LKC, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106990 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-990 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-990 Supplier Abnova Corporation Supplier No. H00054825A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PC-LKC.

This gene is a member of the protocadherin family, which represents a subset of the larger cadherin superfamily. The members of the protocadherin family encode nonclassical cadherins that function as calcium-dependent cell-cell adhesion molecules. The kidney-expressed gene product has a signal peptide, nine cadherin repeat domains and a unique cytoplasmic region. This protocadherin represents a new candidate for tumor suppression. [provided by RefSeq

Sequence: GGMYHNTFTIQCSLPVFLSISVVDQPDLDPQFVREFYSASVAEDAAKGTSVLTVEAVDGDKGINDPVIYSISYSTRPGWFDIGADGVIRVNGSLDREQLLEADEEVQLQ

Specifications

Antigen PC-LKC
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PC-LKC.
Formulation 50% glycerol
Gene PCDH24
Gene Accession No. NM_017675
Gene Alias FLJ20124/FLJ20383/MGC163154/PC-LKC/PCLKC
Gene Symbols PCDH24
Host Species Mouse
Immunogen PC-LKC (NP_060145, 210 a.a. ∽ 318 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54825
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.