Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCBP4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107167 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-167 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-167 Supplier Abnova Corporation Supplier No. H00057060A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PCBP4.

This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene is induced by the p53 tumor suppressor, and the encoded protein can suppress cell proliferation by inducing apoptosis and cell cycle arrest in G(2)-M. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Multiple alternatively spliced transcript variants have been described, but the full-length nature for only some has been determined. [provided by RefSeq]

Sequence: TTGAQVQVAGDLLPNSTERAVTVSGVPDAIILCVRQICAVILESPPKGATIPYHPSLSLGTVLLSANQGFSVQGQYGAVTPAEVTKLQQLSSHAVPFATPSVVPGLDPGT

Specifications

Antigen PCBP4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PCBP4.
Formulation 50% glycerol
Gene PCBP4
Gene Accession No. NM_033010
Gene Alias LIP4/MCG10
Gene Symbols PCBP4
Host Species Mouse
Immunogen PCBP4 (NP_127503, 130 a.a. ∽ 239 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57060
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.