Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH10, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107201 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-201 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-201 Supplier Abnova Corporation Supplier No. H00057575A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PCDH10.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains. [provided by RefSeq

Sequence: SQLHYTVQEEQEHGTFVGNIAEDLGLDITKLSARGFQTVPNSRTPYLDLNLETGVLYVNEKIDREQICKQSPSCVLHLEVFLENPLELFQVEIEVLDINDNPPSFPEPDL

Specifications

Antigen PCDH10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PCDH10.
Formulation 50% glycerol
Gene PCDH10
Gene Accession No. NM_020815
Gene Alias DKFZp761O2023/KIAA1400/MGC133344/OL-PCDH/PCDH19
Gene Symbols PCDH10
Host Species Mouse
Immunogen PCDH10 (NP_065866, 18 a.a. ∽ 127 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57575
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.